Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Phenoxybenzaldehyde, 98%, Thermo Scientific™

Catalog No. AAA1378030
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1378030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1378030 Supplier Thermo Scientific Chemicals Supplier No. A1378030

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 39515-51-0
Melting Point 13°C
Density 1.15
Boiling Point 139°C to 140°C (0.1 mmHg)
Flash Point 186°C (366°F)
Molecular Formula C13H10O2
Refractive Index 1.598
Quantity 250 g
MDL Number MFCD00003353
UN Number UN2810
Beilstein 511662
Sensitivity Air Sensitive
Synonym m-phenoxybenzaldehyde, 3-phenoxy-benzaldehyde, benzaldehyde, 3-phenoxy, 3-formyldiphenyl ether, m-phenyloxy benzaldehyde, 3-phenoxy benzaldehyde, dsstox_cid_8005, dsstox_rid_78636, dsstox_gsid_28005, 5-phenoxybenzaldehyde
InChI Key MRLGCTNJRREZHZ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)C=O
IUPAC Name 3-phenoxybenzaldehyde
Molecular Weight (g/mol) 198.221
PubChem CID 38284
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 3-Phenoxybenzaldehyde
Show More Show Less
Hazard Category H302-H330
Hazard Statement GHS H Statement
H301-H330
Toxic if swallowed.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P312-P304+P340-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 254-487-1
RTECSNumber CU7560200
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.