accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3959-05-5

IUPAC Name: 1-(2-bromophenyl)methanamine
Synonyms: 1-(2-bromophenyl)methanamine
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: NOYASZMZIBFFNZ-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1Br
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results