accessibility menu, dialog, popup

UserName

Viewing profile as user:
FF

CAS RN 396-64-5

IUPAC Name: 3,3'-difluoro-1,1'-biphenyl
Synonyms: 3,3'-difluoro-1,1'-biphenyl
Molecular Weight (g/mol): 190.19
Molecular Formula: C12H8F2
InChi Key: GAYJHUJLHJWCTH-UHFFFAOYSA-N
SMILES: FC1=CC(=CC=C1)C1=CC(F)=CC=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)