Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Difluorobiphenyl 98.0+%, TCI America™
SDP

Catalog No. D21801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21801G Supplier TCI America Supplier No. D21801G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 396-64-5
Molecular Formula C12H8F2
Molecular Weight (g/mol) 190.19
MDL Number MFCD00039216
InChI Key GAYJHUJLHJWCTH-UHFFFAOYSA-N
Synonym 3,3'-difluorobiphenyl, 3,3'-difluoro-1,1'-biphenyl, 1-fluoro-3-3-fluorophenyl benzene, 1,1'-biphenyl,3,3'-difluoro, 1,1'-biphenyl, 3,3'-difluoro, 3-fluoro-1-3-fluorophenyl benzene, acmc-1agdg, 3,3'-difluoro biphenyl, gayjhujlhjwcth-uhfffaoysa, 3,3'-difluoro-1,1'-biphenyl #
PubChem CID 123058
IUPAC Name 3,3'-difluoro-1,1'-biphenyl
SMILES FC1=CC(=CC=C1)C1=CC(F)=CC=C1

Specifications

Color Yellow
Boiling Point 130°C
Quantity 1 g
UN Number 3151
Formula Weight 190.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3′-Difluorobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.