accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 396129-53-6

IUPAC Name: 4-[5-(pyridin-2-yl)-1H-pyrazol-4-yl]quinoline
Synonyms: 4-[3-(pyridin-2-yl)-2H-pyrazol-4-yl]quinoline
Molecular Weight (g/mol): 272.31
Molecular Formula: C17H12N4
InChi Key: IBCXZJCWDGCXQT-UHFFFAOYSA-N
SMILES: N1N=CC(=C1C1=CC=CC=N1)C1=CC=NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results