accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 396131-82-1

BOOH3CCH3H3CH3CN

CAS RN 396131-82-1

IUPAC Name: 2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
Synonyms: 2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
Molecular Weight (g/mol): 243.11
Molecular Formula: C14H18BNO2
InChi Key: LIWPKYSGWSFPEE-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(CC#N)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results