Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Cyanomethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH5261806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5261806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5261806 Supplier Thermo Scientific Chemicals Supplier No. H5261806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(Cyanomethyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 396131-82-1
Molecular Formula C14H18BNO2
Molecular Weight (g/mol) 243.11
MDL Number MFCD02179458
InChI Key LIWPKYSGWSFPEE-UHFFFAOYSA-N
Synonym 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-cyanomethyl phenylboronic acid pinacol ester, 2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, pubchem7790, 3-cyanomethylphenylboronic acid, pinacol ester, 3-cyanomethylphenyl boronic acid,pinacol ester, 3-cyanomethyl phenylboronic acid, pinacol ester
PubChem CID 2773343
IUPAC Name 2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CC(CC#N)=CC=C1

Specifications

Melting Point 44°C to 48°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 243.11
Percent Purity 96%
Chemical Name or Material 3-(Cyanomethyl)benzeneboronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.