accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3963-95-9

IUPAC Name: hydrogen (2Z,4S,4aR,5S,5aR,12aS)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-5,10,11,12a-tetrahydroxy-6-methylidene-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Synonyms: hydrogen methacyclin chloride
Molecular Weight (g/mol): 478.88
Molecular Formula: C22H23ClN2O8
InChi Key: MIVNJKLNBAONDJ-MCKSDNNQSA-N
SMILES: [H+].[Cl-].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@@H]3C(=C)C4=CC=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
Grade 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results