Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Metacycline hydrochloride, 98%, sec. Standard

Catalog No. AC303861000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303861000 100 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC303861000 Supplier Thermo Scientific Chemicals Supplier No. 303861000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3963-95-9
Molecular Formula C22H22N2O8·HCl
Molecular Weight (g/mol) 478.9
MDL Number MFCD04972012
InChI Key VXPSARQTYDZXAO-CCHMMTNSSA-N
Synonym methacycline hydrochloride, methacycline hcl, adriamicina, ciclobiotic, germiciclin, rondomycin, metadomus, metacycline hydrochloride, globociclina, metilenbiotic
PubChem CID 54685047
IUPAC Name (4S,4aR,5S,5aR,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methylidene-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CN(C)C1C2C(C3C(=C)C4=C(C(=CC=C4)O)C(=C3C(=O)C2(C(=C(C1=O)C(=O)N)O)O)O)O.Cl

Specifications

Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 100 mg
Merck Index 15, 6015
Formula Weight 478.9
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Metacycline hydrochloride, 98%
Hazard Category LIGHT SENSITIVE
EINECSNumber 223-568-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.