accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOOOCl

CAS RN 3967-55-3

IUPAC Name: 4,5-dichloro-1,3-dioxolan-2-one
Synonyms: 4,5-dichloro-1,3-dioxolan-2-one
Molecular Weight (g/mol): 156.95
Molecular Formula: C3H2Cl2O3
InChi Key: BETICXVUVYXEJX-UHFFFAOYNA-N
SMILES: ClC1OC(=O)OC1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results