Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dichloro-1,3-dioxolan-2-one, MP Biomedicals

Catalog No. ICN201113 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN201113 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN201113 Supplier MP Biomedicals, Inc Supplier No. 0520111310

Chemical Identifiers

CAS 3967-55-3
Molecular Formula C3H2Cl2O3
Molecular Weight (g/mol) 156.946
InChI Key BETICXVUVYXEJX-UHFFFAOYSA-N
Synonym cyclic 1,2-dichloroethylene carbonate, 1,2-dichloroethylene carbonate, 4,5-dichloro-1,3-dioxolane-2-one, 1,3-dioxolane, 4,5-dichloro-2-oxo, carbonic acid, cyclic 1,2-dichloroethylene ester, 1,3-dioxolan-2-one, 4,5-dichloro, 1,2-dichloro ethylene carbonate, 4,3-dioxolane-2-one, dichloroethylene carbonate, carbonic acid, 1,2-dichloroethylene ester 6ci
PubChem CID 19869
IUPAC Name 4,5-dichloro-1,3-dioxolan-2-one
SMILES C1(C(OC(=O)O1)Cl)Cl

Specifications

Quantity 10 g
Formula Weight 156.95
Chemical Name or Material 4, 5-Dichloro-1, 3-dioxolan-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.