accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39691-62-8

IUPAC Name: bromo(nonyl)magnesium
Synonyms: bromo(nonyl)magnesium
Molecular Weight (g/mol): 231.46
Molecular Formula: C9H19BrMg
InChi Key: IFQPKTPQJLAZHQ-UHFFFAOYSA-M
SMILES: CCCCCCCCC[Mg]Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results