Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™

Catalog No. AC440781000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC440781000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC440781000 Supplier Thermo Scientific Chemicals Supplier No. 440781000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 39691-62-8 | C9H19BrMg | 231.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39691-62-8 , 60-29-7
Molecular Formula C9H19BrMg
Molecular Weight (g/mol) 231.46
MDL Number MFCD03094048
InChI Key IFQPKTPQJLAZHQ-UHFFFAOYSA-M
Synonym nonylmagnesium bromide, nonylmagnesium bromide solution, ifqpktpqjlazhq-uhfffaoysa-m, nonylmagnesium bromide 1.0m solution i, nonylmagnesium bromide, 0.5m in tetrahydrofuran, nonylmagnesium bromide 1.0m solution i, nonylmagnesium bromide solution, 1.0 m in diethyl ether, grignard reagent
PubChem CID 11042600
IUPAC Name magnesium;nonane;bromide
SMILES CCCCCCCCC[Mg]Br

Specifications

Color Colorless to Brown
Density 0.8490g/mL
Boiling Point 35.0°C
Flash Point −30°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3(CH2)8MgBr
Quantity 100 mL
Specific Gravity 0.849
Solubility Information Solubility in water: reacts
Formula Weight 231.46
Physical Form Liquid
Chemical Name or Material Nonylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Reacts violently with water.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.