accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COOHNHOOH

CAS RN 397246-14-9

IUPAC Name: tert-butyl N-(1,4-dihydroxybutan-2-yl)carbamate
Synonyms: tert-butyl N-(1,4-dihydroxybutan-2-yl)carbamate
Molecular Weight (g/mol): 205.25
Molecular Formula: C9H19NO4
InChi Key: KLRRFBSWOIUAHZ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CO)CCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results