accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39794-75-7

IUPAC Name: ethyl 2-[(4-methylbenzenesulfonyl)oxy]acetate
Synonyms: ethyl 2-[(4-methylbenzenesulfonyl)oxy]acetate
Molecular Weight (g/mol): 258.29
Molecular Formula: C11H14O5S
InChi Key: CVNRARCXNKDWKI-UHFFFAOYSA-N
SMILES: CCOC(=O)COS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results