accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 39815-78-6

IUPAC Name: methyl 3-oxoheptanoate
Synonyms: methyl 3-oxoheptanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: CZTKGERSDUGZPQ-UHFFFAOYSA-N
SMILES: CCCCC(=O)CC(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results