Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-Oxoheptanoate 95.0+%, TCI America™
SDP

Catalog No. O024625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O024625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O024625ML Supplier TCI America Supplier No. O024625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39815-78-6
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00191568
InChI Key CZTKGERSDUGZPQ-UHFFFAOYSA-N
Synonym 3-Ketoheptanoic Acid Methyl Ester, Methyl 3-Oxoenanthate, 3-Oxoenanthic Acid Methyl Ester, 3-Oxoheptanoic Acid Methyl Ester
PubChem CID 546075
IUPAC Name methyl 3-oxoheptanoate
SMILES CCCCC(=O)CC(=O)OC

Specifications

Color Yellow
Boiling Point 95°C
Quantity 25 mL
Formula Weight 158.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Oxoheptanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.