accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2BrBr

CAS RN 39856-57-0

IUPAC Name: 2,6-dibromopyridin-3-amine
Synonyms: 2,6-dibromopyridin-3-amine
Molecular Weight (g/mol): 251.91
Molecular Formula: C5H4Br2N2
InChi Key: CRTOIQFRVBJJRI-UHFFFAOYSA-N
SMILES: NC1=C(Br)N=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results