Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2,6-dibromopyridine 98.0+%, TCI America™
SDP

Catalog No. A22725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22725G 5 g
A22721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22725G Supplier TCI America Supplier No. A22725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 39856-57-0 | C5H4Br2N2 | 251.909 g/mol

Chemical Identifiers

CAS 39856-57-0
Molecular Formula C5H4Br2N2
Molecular Weight (g/mol) 251.909
MDL Number MFCD00128862
InChI Key CRTOIQFRVBJJRI-UHFFFAOYSA-N
Synonym 3-amino-2,6-dibromopyridine, 2,6-dibromo-3-pyridinamine, 2,6-dibromo-pyridin-3-ylamine, 3-pyridinamine, 2,6-dibromo, 2,6-dibromo-3-pyridylamine, 2,6-dibromo-3-aminopyridine, zlchem 949, pubchem9809, acmc-209j8e, ksc495i3f
PubChem CID 817678
IUPAC Name 2,6-dibromopyridin-3-amine
SMILES C1=CC(=NC(=C1N)Br)Br

Specifications

Melting Point 147°C
Quantity 5 g
Formula Weight 251.91
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2,6-dibromopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.