accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 39859-36-4

IUPAC Name: 2-benzoyl-4-bromoaniline
Synonyms: 2-benzoyl-4-bromoaniline
Molecular Weight (g/mol): 276.13
Molecular Formula: C13H10BrNO
InChi Key: LXJVUGANBDAASB-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C=C1C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results