accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3Br

CAS RN 3989-13-7

IUPAC Name: [2-(3-bromophenyl)ethynyl]trimethylsilane
Synonyms: [2-(3-bromophenyl)ethynyl]trimethylsilane
Molecular Weight (g/mol): 253.21
Molecular Formula: C11H13BrSi
InChi Key: BXXMHQYHYOTTPM-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC(Br)=CC=C1
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)