accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 399-10-0

IUPAC Name: (2E)-1-(4-fluorophenyl)-3-phenylprop-2-en-1-one
Synonyms: (2E)-1-(4-fluorophenyl)-3-phenylprop-2-en-1-one
Molecular Weight (g/mol): 226.25
Molecular Formula: C15H11FO
InChi Key: VKNQSJQWRINEFS-IZZDOVSWSA-N
SMILES: FC1=CC=C(C=C1)C(=O)\C=C\C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results