accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClNHNH2

CAS RN 39943-56-1

IUPAC Name: (3,5-dichlorophenyl)hydrazine
Synonyms: (3,5-dichlorophenyl)hydrazine
Molecular Weight (g/mol): 177.03
Molecular Formula: C6H6Cl2N2
InChi Key: ZQOCIFROLPHOEF-UHFFFAOYSA-N
SMILES: NNC1=CC(Cl)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results