accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)NOO

CAS RN 39945-51-2

IUPAC Name: benzyl (3R)-3-(hydroxymethyl)piperidine-1-carboxylate
Synonyms: benzyl (3R)-3-(hydroxymethyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 249.31
Molecular Formula: C14H19NO3
InChi Key: XLWOOUZKMJBINO-CYBMUJFWSA-N
SMILES: OC[C@@H]1CCCN(C1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results