accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOCH2HO

CAS RN 400091-05-6

IUPAC Name: 6-(4-methoxyphenoxy)-2-phenyl-8-(prop-2-en-1-yloxy)-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-ol
Synonyms: 6-(4-methoxyphenoxy)-2-phenyl-8-(prop-2-en-1-yloxy)-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-ol
Molecular Weight (g/mol): 414.45
Molecular Formula: C23H26O7
InChi Key: BALLDQXJTPWKTQ-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC3COC(OC3C(OCC=C)C2O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results