accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NCS

CAS RN 40046-30-8

IUPAC Name: 1-isothiocyanato-3,5-dimethylbenzene
Synonyms: 1-isothiocyanato-3,5-dimethylbenzene
Molecular Weight (g/mol): 163.24
Molecular Formula: C9H9NS
InChi Key: DSMXCADWIFIJEX-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)N=C=S
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)