Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylphenyl Isothiocyanate 97.0+%, TCI America™
SDP

Catalog No. D49445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49445G 5 g
D494425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49445G Supplier TCI America Supplier No. D49445G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 40046-30-8 | C9H9NS | 163.238 g/mol

Chemical Identifiers

CAS 40046-30-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
MDL Number MFCD00041082
InChI Key DSMXCADWIFIJEX-UHFFFAOYSA-N
Synonym 3,5-dimethylphenyl isothiocyanate, 3,5-dimethylphenylisothiocyanate, 3,5-dimethylbenzenisothiocyanate, acmc-20anca, 3,5-xylyl isothiocyanate, 1-isothiocyanato-3,5-dimethyl-benzene, #, isothiocyanic acid 3,5-dimethylphenyl ester
PubChem CID 142406
IUPAC Name 1-isothiocyanato-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)N=C=S)C

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 2810
Boiling Point 140°C
Formula Weight 163.24
Percent Purity ≥97.0% (GC)
Grade Not Available
Chemical Name or Material 3,5-Dimethylphenyl Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.