accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 40061-55-0

IUPAC Name: ethyl 2-(3-methylphenyl)acetate
Synonyms: ethyl 2-(3-methylphenyl)acetate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: RVIWPVAVHHVQIR-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results