Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl m-Tolylacetate 98.0+%, TCI America™
SDP

Catalog No. T122610G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T122610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T122610G Supplier TCI America Supplier No. T122610G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40061-55-0
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00017273
InChI Key RVIWPVAVHHVQIR-UHFFFAOYSA-N
Synonym ethyl m-tolylacetate, ethyl 2-3-methylphenyl acetate, ethyl meta-tolylacetate, ethyl 3-methylphenylacetate, m-tolylacetic acid ethyl ester, ethyl 2-m-tolyl acetate, benzeneacetic acid, 3-methyl-, ethyl ester, ethyl m-methylphenylacetate, 3-methylphenylacetic acid ethyl ester, ethylm-tolylacetate
PubChem CID 96574
IUPAC Name ethyl 2-(3-methylphenyl)acetate
SMILES CCOC(=O)CC1=CC=CC(=C1)C

Specifications

Color Yellow
Boiling Point 238°C
Quantity 10 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl m-Tolylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.