accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4007-01-6

IUPAC Name: 2-[(dimethylcarbamothioyl)sulfanyl]acetic acid
Synonyms: [(dimethylcarbamothioyl)sulfanyl]acetic acid
Molecular Weight (g/mol): 179.25
Molecular Formula: C5H9NO2S2
InChi Key: FYGUGMCPHDSCLF-UHFFFAOYSA-N
SMILES: CN(C)C(=S)SCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results