Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-(N,N-Dimethylthiocarbamoyl)thioglycolic Acid 97.0+%, TCI America™
SDP

Catalog No. D25005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25005G Supplier TCI America Supplier No. D25005G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4007-01-6
Molecular Formula C5H9NO2S2
Molecular Weight (g/mol) 179.25
MDL Number MFCD00021765
InChI Key FYGUGMCPHDSCLF-UHFFFAOYSA-N
Synonym S-(N,N-Dimethylthiocarbamoyl)mercaptoacetic Acid
PubChem CID 301695
IUPAC Name 2-(dimethylcarbamothioylsulfanyl)acetic acid
SMILES CN(C)C(=S)SCC(=O)O

Specifications

Melting Point 150°C
Color White
Quantity 5 g
Formula Weight 179.25
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material S-(N,N-Dimethylthiocarbamoyl)thioglycolic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.