accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40070-59-5

IUPAC Name: (3S)-3-(3-bromo-4-hydroxy-2,5-dimethylphenyl)-3-[(1S,4S,5R,6S)-5-bromo-4-hydroxy-3,6-dimethylcyclohex-2-en-1-yl]-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: (3S)-3-(3-bromo-4-hydroxy-2,5-dimethylphenyl)-3-[(1S,4S,5R,6S)-5-bromo-4-hydroxy-3,6-dimethylcyclohex-2-en-1-yl]-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Weight (g/mol): 572.31
Molecular Formula: C23H24Br2O5S
InChi Key: JHWIBPVPGLHJHA-RMFGFXDFSA-N
SMILES: C[C@@H]1[C@@H](Br)[C@@H](O)C(C)=C[C@@H]1[C@]1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1C
Quantity 
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results