accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHONCH3OCH3

CAS RN 400898-92-2

IUPAC Name: tert-butyl N-{4-[methoxy(methyl)carbamoyl]cyclohexyl}carbamate
Synonyms: tert-butyl N-{4-[methoxy(methyl)carbamoyl]cyclohexyl}carbamate
Molecular Weight (g/mol): 286.37
Molecular Formula: C14H26N2O4
InChi Key: VCYMEIIREZLGFN-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C1CCC(CC1)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)