Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1-(Boc-amino)-4-(N-methoxy-N-methylcarbamoyl)cyclohexane, 97%

Catalog No. AAH6239706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6239706 5 g
AAH6239703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6239706 Supplier Thermo Scientific Chemicals Supplier No. H6239706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 400898-92-2 | C14H26N2O4 | 286.372 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 400898-92-2
Molecular Formula C14H26N2O4
Molecular Weight (g/mol) 286.372
MDL Number MFCD08436075
InChI Key VCYMEIIREZLGFN-UHFFFAOYSA-N
Synonym tert-butyl cis-4-n-methoxy-n-methylcarbamoyl-cyclohexylcarbamate, tert-butyl trans-4-methoxy methyl carbamoyl cyclohexyl carbamate, tert-butyl trans-4-n-methoxy-n-methylcarbamoyl cyclohexylcarbamate, trans-1-boc-amino-4-n-methoxy-n-methylcarbamoyl cyclohexane, vcymeiirezlgfn-phimtyicsa-n, vcymeiirezlgfn-xypyzodxsa-n, tert-butyl 4-methoxy methyl carbamoyl cyclohexyl carbamate, tert-butyl cis-4-methoxy methyl carbamoyl cyclohexylcarbamate, tert-butyl cis-4-methoxy methyl carbamoyl cyclohexyl carbamate
PubChem CID 24720928
IUPAC Name tert-butyl N-[4-[methoxy(methyl)carbamoyl]cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C(=O)N(C)OC

Specifications

Quantity 5 g
Formula Weight 286.36
Percent Purity 97%
Chemical Name or Material trans-1-(Boc-amino)-4-(N-methoxy-N-methylcarbamoyl)cyclohexane
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.