CAS RN 401-67-2
IUPAC Name:
2,2-difluoroacetyl 2,2-difluoroacetate
Synonyms:
2,2-difluoroacetyl 2,2-difluoroacetate
Molecular Weight (g/mol):
174.05
Molecular Formula:
C4H2F4O3
InChi Key:
IYXUFOCLMOXQSL-UHFFFAOYSA-N
SMILES:
FC(F)C(=O)OC(=O)C(F)F
1
–
1
of
1
results
1
Difluoroacetic Anhydride 98.0+%, TCI America™
CAS: 401-67-2 Molecular Formula: C4H2F4O3 Molecular Weight (g/mol): 174.051 MDL Number: MFCD02093315 InChI Key: IYXUFOCLMOXQSL-UHFFFAOYSA-N PubChem CID: 2774048 IUPAC Name: (2,2-difluoroacetyl) 2,2-difluoroacetate SMILES: C(C(=O)OC(=O)C(F)F)(F)F