Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Difluoroacetic Anhydride 98.0+%, TCI America™
SDP

Catalog No. D41645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41645G Supplier TCI America Supplier No. D41645G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 401-67-2
Molecular Formula C4H2F4O3
Molecular Weight (g/mol) 174.051
MDL Number MFCD02093315
InChI Key IYXUFOCLMOXQSL-UHFFFAOYSA-N
PubChem CID 2774048
IUPAC Name (2,2-difluoroacetyl) 2,2-difluoroacetate
SMILES C(C(=O)OC(=O)C(F)F)(F)F

Specifications

Boiling Point 127°C
Quantity 5 g
UN Number 3265
Formula Weight 174.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Difluoroacetic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.