accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OO

CAS RN 4016-14-2

IUPAC Name: 2-[(propan-2-yloxy)methyl]oxirane
Synonyms: isopropyl glycidyl ether
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: NWLUZGJDEZBBRH-UHFFFAOYNA-N
SMILES: CC(C)OCC1CO1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)