Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycidyl Isopropyl Ether 96.0+%, TCI America™
SDP

Catalog No. G0255500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0255500ML 500 mL
G025525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0255500ML Supplier TCI America Supplier No. G0255500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4016-14-2
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00005139
InChI Key NWLUZGJDEZBBRH-UHFFFAOYSA-N
Synonym glycidyl isopropyl ether, isopropyl glycidyl ether, 2,3-epoxypropyl isopropyl ether, isopropoxymethyl oxirane, 2-isopropoxymethyl oxirane, 3-isopropyloxypropylene oxide, 3-isopropoxy-1,2-epoxypropane, isopropyl epoxypropyl ether, propane, 1,2-epoxy-3-isopropoxy, oxirane, 1-methylethoxy methyl
PubChem CID 19920
IUPAC Name 2-(propan-2-yloxymethyl)oxirane
SMILES CC(C)OCC1CO1

Specifications

Color Colorless
Boiling Point 132°C
Quantity 500 mL
UN Number 3271
Formula Weight 116.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycidyl Isopropyl Ether
RTECSNumber TZ3500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.