accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4016-63-1

IUPAC Name: 2-amino-8-bromo-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Synonyms: 8-bromoguanosine
Molecular Weight (g/mol): 362.14
Molecular Formula: C10H12BrN5O5
InChi Key: ASUCSHXLTWZYBA-JJJZDVOVNA-N
SMILES: NC1=NC2=C(N=C(Br)N2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results