accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40172-95-0

IUPAC Name: 4-(furan-2-carbonyl)piperazin-1-ium
Synonyms: 4-(furan-2-carbonyl)piperazin-1-ium
Molecular Weight (g/mol): 181.21
Molecular Formula: C9H13N2O2
InChi Key: SADPINFEWFPMEA-UHFFFAOYSA-O
SMILES: O=C(N1CC[NH2+]CC1)C1=CC=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results