Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Furoyl)piperazine, 98%, Thermo Scientific™

Catalog No. AAA1677322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1677322 100 g
AAA1677314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1677322 Supplier Thermo Scientific Chemicals Supplier No. A1677322

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40172-95-0
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.207
MDL Number MFCD00038831
InChI Key SADPINFEWFPMEA-UHFFFAOYSA-N
Synonym 1-2-furoyl piperazine, 1-furan-2-carbonyl piperazine, furan-2-yl piperazin-1-yl methanone, n-2-furoyl piperazine, unii-8aux72ll1p, 1-2-furoyl-piperazine, furan-2-yl-piperazin-1-yl-methanone, piperazine, 1-2-furanylcarbonyl, 8aux72ll1p
PubChem CID 550206
IUPAC Name furan-2-yl(piperazin-1-yl)methanone
SMILES C1CN(CCN1)C(=O)C2=CC=CO2

Specifications

Melting Point 66°C to 70°C
Boiling Point 137°C to 140°C (0.1 mmHg)
Quantity 100 g
Beilstein 880196
Sensitivity Air Sensitive
Formula Weight 180.21
Percent Purity 98%
Chemical Name or Material 1-(2-Furoyl)piperazine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 254-823-7
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.