accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CCH3CH3

CAS RN 401797-00-0

IUPAC Name: 2-(3,4-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3,4-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 232.13
Molecular Formula: C14H21BO2
InChi Key: HWIMTTLCFONAAB-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C(C=C1)B1OC(C)(C)C(C)(C)O1
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results