Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
SDP

Catalog No. D50851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D50851G Supplier TCI America Supplier No. D50851G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 401797-00-0 | C14H21BO2 | 232.13 g/mol

Chemical Identifiers

CAS 401797-00-0
Molecular Formula C14H21BO2
Molecular Weight (g/mol) 232.13
MDL Number MFCD12405519
InChI Key HWIMTTLCFONAAB-UHFFFAOYSA-N
Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-o-xylene, 3,4-Dimethylphenylboronic Acid Pinacol Ester
PubChem CID 15906194
IUPAC Name 2-(3,4-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C)C

Specifications

Color White
Quantity 1 g
Physical Form Crystal-Lump at 20°C
Formula Weight 232.13
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Chemical Name or Material 2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.