accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4018-65-9

IUPAC Name: 3-chlorobenzene-1,2-diol
Synonyms: 3-chlorocatechol
Molecular Weight (g/mol): 144.55
Molecular Formula: C6H5ClO2
InChi Key: GQKDZDYQXPOXEM-UHFFFAOYSA-N
SMILES: OC1=CC=CC(Cl)=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results