Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorocatechol 98.0+%, TCI America™
SDP

Catalog No. C12461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C12461G Supplier TCI America Supplier No. C12461G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4018-65-9
Molecular Formula C6H5ClO2
Molecular Weight (g/mol) 144.554
MDL Number MFCD00143111
InChI Key GQKDZDYQXPOXEM-UHFFFAOYSA-N
Synonym 1-Chloro-2,3-dihydroxybenzene
PubChem CID 19928
ChEBI CHEBI:27715
IUPAC Name 3-chlorobenzene-1,2-diol
SMILES C1=CC(=C(C(=C1)Cl)O)O

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 138°C
Quantity 1 g
Formula Weight 144.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chlorocatechol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.