accessibility menu, dialog, popup

UserName

Viewing profile as user:
SFFFFFO

CAS RN 401892-80-6

IUPAC Name: 3-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Synonyms: 3-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Molecular Weight (g/mol): 232.17
Molecular Formula: C7H5F5OS
InChi Key: APFMXGBPENUHDD-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results