accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 402-41-5

IUPAC Name: 2-(4-fluorophenyl)propan-2-ol
Synonyms: 2-(4-fluorophenyl)propan-2-ol
Molecular Weight (g/mol): 154.18
Molecular Formula: C9H11FO
InChi Key: QURXIISLVHJNGB-UHFFFAOYSA-N
SMILES: CC(C)(O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results