Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Fluorophenyl)-2-propanol 95.0+%, TCI America™
SDP

Catalog No. F024210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F024210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F024210G Supplier TCI America Supplier No. F024210G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 402-41-5
Molecular Formula C9H11FO
Molecular Weight (g/mol) 154.184
MDL Number MFCD00059919
InChI Key QURXIISLVHJNGB-UHFFFAOYSA-N
PubChem CID 637733
IUPAC Name 2-(4-fluorophenyl)propan-2-ol
SMILES CC(C)(C1=CC=C(C=C1)F)O

Specifications

Color Yellow
Boiling Point 61°C
Quantity 10 g
Formula Weight 154.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Fluorophenyl)-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.