accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONF

CAS RN 402-66-4

IUPAC Name: 5-fluoropyridine-3-carboxylic acid
Synonyms: 5-fluoropyridine-3-carboxylic acid
Molecular Weight (g/mol): 141.1
Molecular Formula: C6H4FNO2
InChi Key: BXZSBDDOYIWMGC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=CC(F)=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)