Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoronicotinic acid, 98%

Catalog No. AAH6116222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6116222 100 g
AAH6116206 5 g
AAH6116214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6116222 Supplier Thermo Scientific Chemicals Supplier No. H6116222

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

CAS: 402-66-4 | C6H4FNO2 | 141.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-66-4
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD01318537
InChI Key BXZSBDDOYIWMGC-UHFFFAOYSA-N
Synonym 5-fluoronicotinic acid, 5-fluoro-3-pyridinecarboxylic acid, 5-fluoro-nicotinic acid, 3-pyridinecarboxylic acid, 5-fluoro, nicotinic acid, 5-fluoro, 3-fluoro-5-carboxypyridine, 3-fluoro-5-pyridinecarboxylic acid, 5-fluoronicotinicacid, pubchem10942
PubChem CID 120232
IUPAC Name 5-fluoropyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=CC(F)=C1

Specifications

Melting Point 193°C to 198°C
Color White
Quantity 100 g
Solubility Information Soluble in methanol.
Formula Weight 141.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Fluoronicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber US5745250
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.